(R,R)-1,2-Bis[(2-methylphenyl)-(phenyl)phosphino]ethane ≥95%

Supplier: Matrix Scientific

115242-500MG 115242-1G
102893-032EA 1340 USD
102893-032 102893-034
(R,R)-1,2-Bis[(2-methylphenyl)-(phenyl)phosphino]ethane ≥95%
(R,R)-1,2-Bis[(2-methylphenyl)-(phenyl)phosphino]ethane

Hazard: Irritant

Formula: C₂₈H₂₈P₂
MW: 426.48 g/mol
MDL Number: MFCD12198305
CAS Number: 81157-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR