(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2-Bis((S)-(2-methoxyphenyl)(phenyl)phosphino)ethane, (S,S)-Ethylenebis[(2-methoxyphenyl)phenylphosphine], (S,S)-(+)-1,2-Bis[(2-methoxyphenyl)(phenyl)phosphino]ethane, (+)-DIPAMP, (S,S)-DIPAMP

115241-500MG 115241-1G
102893-028EA 804.62 USD
102893-028 102893-030
(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane ≥95%
(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane

Hazard: Irritant

Formula: C₂₈H₂₈O₂P₂
MW: 458.48 g/mol
Melting Pt: 101…105 °C
MDL Number: MFCD05863547
CAS Number: 97858-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR