5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Amino-7-bromo-9-methyl-7-deazapurine

115236-1G 115236-500MG
102893-016EA 564.62 USD
102893-016 102895-670
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine ≥97%
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

Hazard: Irritant

Formula: C₇H₇BrN₄
MW: 227.065 g/mol
MDL Number: MFCD20486185
CAS Number: 1337532-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR