1-(4-Nitrophenyl)-4-(trifluoromethyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

115233-500MG 115233-1G
102895-662EA 836.92 USD
102895-662 102895-664
1-(4-Nitrophenyl)-4-(trifluoromethyl)-1H-pyrazole ≥95%
1-(4-Nitrophenyl)-4-(trifluoromethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₆F₃N₃O₂
Melting Pt: 93…95 °C
MDL Number: MFCD28023542
CAS Number: 1393125-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR