Sodium-3-hydroxynaphthalene-2-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Hydroxy-3-naphthoic acid sodium salt, Sodium 3-Hydroxy-2-naphthoate, 3-Hydroxy-2-naphthoic acid sodium salt
115226-500MG
115226-1G
102892-582EA
75.38
USD
102892-582
102895-636
Sodium-3-hydroxynaphthalene-2-carboxylate ≥97%
Sodium-3-hydroxynaphthalene-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₇NaO₃ MW: 210.17 g/mol |
MDL Number:
MFCD00068251 CAS Number: 14206-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...