2-Hydroxy-4-(methylsulfanyl)benzoic acid ≥97%

Supplier: Matrix Scientific

115224-500MG 115224-1G
102892-574EA 1040 USD
102892-574 102892-576
2-Hydroxy-4-(methylsulfanyl)benzoic acid ≥97%
2-Hydroxy-4-(methylsulfanyl)benzoic acid

Hazard: Irritant

Formula: C₈H₈O₃S
MDL Number: MFCD09787510
CAS Number: 67127-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR