3-Bromo-1-N-methylbenzene-1,2-diamine ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-N1-methylbenzene-1,2-diamine
115222-500MG
115222-1G
102895-630EA
836.92
USD
102895-630
102895-632
3-Bromo-1-N-methylbenzene-1,2-diamine ≥97%
3-Bromo-1-N-methylbenzene-1,2-diamine
Hazard: Irritant
Formula:
C₇H₉BrN₂ MW: 201.07 g/mol Melting Pt: 38 °C |
MDL Number:
MFCD12024390 CAS Number: 1150617-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...