3-Bromo-N-methyl-2-nitroaniline ≥97%

Supplier: Matrix Scientific

115221-500MG 115221-1G
102895-626EA 624.62 USD
102895-626 102895-628
3-Bromo-N-methyl-2-nitroaniline ≥97%
3-Bromo-N-methyl-2-nitroaniline

Hazard: Irritant

Formula: C₇H₇BrN₂O₂
Melting Pt: 50…51 °C
MDL Number: MFCD12024389
CAS Number: 1150617-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR