4-Bromo-2-iodotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-iodo-1-methylbenzene

115214-500MG 115214-1G
102895-606EA 75.38 USD
102895-606 102895-608
4-Bromo-2-iodotoluene ≥97%
4-Bromo-2-iodotoluene

Hazard: Irritant

Formula: C₇H₆BrI
MW: 296.9300 g/mol
MDL Number: MFCD07780671
CAS Number: 260558-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR