2-Bromo-5-iodoanisole ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Bromo-4-iodo-2-methoxybenzene
115211-500MG
115211-1G
115211-5G
102892-416EA
204.62
USD
102892-416
102892-418
102892-420
2-Bromo-5-iodoanisole ≥97%
2-Bromo-5-iodoanisole
Hazard: Irritant
Formula:
C₇H₆BrIO MW: 312.93 g/mol |
MDL Number:
MFCD07780651 CAS Number: 755027-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...