6-Bromo-o-cresol ≥96%

Supplier: Matrix Scientific

115209-1G 115209-5G
102892-410EA 44.62 USD
102892-410 102892-412
6-Bromo-o-cresol ≥96%
6-Bromo-o-cresol

Hazard: Irritant-Harmful/Stored under Argon/Light Sensitive

Formula: C₇H₇BrO
Boiling Pt: 207…210 °C
Melting Pt: 16.5 °C
Density: 1.51
MDL Number: MFCD09907876
CAS Number: 13319-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR