rac-5-(4-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

115207-500MG 115207-1G
102895-602EA 624.62 USD
102895-602 102895-604
rac-5-(4-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid ≥97%
rac-5-(4-Fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₈FNO₂S
MDL Number: MFCD09907872
CAS Number: 433283-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR