1-(Cyclopropylmethyl)piperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

115202-500MG 115202-1G
102895-588EA 423.08 USD
102895-588 102895-590
1-(Cyclopropylmethyl)piperazine dihydrochloride ≥95%
1-(Cyclopropylmethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₈Cl₂N₂
MW: 213.146 g/mol
MDL Number: MFCD08544420
CAS Number: 373608-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR