2-Bromo-3-methoxyaniline hydrochloride ≥95%

Supplier: Matrix Scientific

115201-500MG 115201-1G
102895-584EA 1040 USD
102895-584 102895-586
2-Bromo-3-methoxyaniline hydrochloride ≥95%
2-Bromo-3-methoxyaniline hydrochloride

Hazard: Irritant

Formula: C₇H₉BrClNO
MDL Number: MFCD09907862
CAS Number: 67853-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR