8-Bromo-7-methylquinoline 97

Supplier: Matrix Scientific

115200-500MG 115200-1G
102895-550EA 836.92 USD
102895-550 102895-582
8-Bromo-7-methylquinoline 97
8-Bromo-7-methylquinoline

Hazard: Irritant

Formula: C₁₀H₈BrN
MW: 222.09 g/mol
CAS Number: 98203-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR