8-Bromo-6-methylquinoline ≥97%

Supplier: Matrix Scientific

115199-500MG 115199-1G
102895-546EA 204.62 USD
102895-546 102895-548
8-Bromo-6-methylquinoline ≥97%
8-Bromo-6-methylquinoline

Hazard: Irritant

Formula: C₁₀H₈BrN
MDL Number: MFCD09701455
CAS Number: 84839-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR