6-Fluoro-8-nitroquinoline ≥97%

Supplier: Matrix Scientific

115198-500MG 115198-1G
102895-542EA 836.92 USD
102895-542 102895-544
6-Fluoro-8-nitroquinoline ≥97%
6-Fluoro-8-nitroquinoline

Hazard: Irritant

Formula: C₉H₅FN₂O₂
MDL Number: MFCD09907841
CAS Number: 343-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR