8-Bromo-5-fluoroquinoline ≥97%

Supplier: Matrix Scientific

115197-500MG 115197-1G
102892-354EA 312.31 USD
102892-354 102895-540
8-Bromo-5-fluoroquinoline ≥97%
8-Bromo-5-fluoroquinoline

Hazard: Irritant

Formula: C₉H₅BrFN
MW: 226.0451 g/mol
MDL Number: MFCD09907839
CAS Number: 917251-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR