8-Bromo-6-fluoroquinoline ≥97%
Supplier: Matrix Scientific
115196-500MG
115196-1G
102892-350EA
312.31
USD
102892-350
102892-352
8-Bromo-6-fluoroquinoline ≥97%
8-Bromo-6-fluoroquinoline
Hazard: Irritant
Formula:
C₉H₅BrFN MW: 226.0451 g/mol |
MDL Number:
MFCD09907837 CAS Number: 22960-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...