2-(4-Bromophenyl)-1,3-oxazole ≥90%

Supplier: Matrix Scientific

115193-500MG 115193-1G
102892-338EA 1040 USD
102892-338 102892-340
2-(4-Bromophenyl)-1,3-oxazole ≥90%
2-(4-Bromophenyl)-1,3-oxazole

Hazard: Irritant

Formula: C₉H₆BrNO
MDL Number: MFCD04973403
CAS Number: 176961-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR