4-Chloro-N-(2,5-difluorophenyl)benzenesulfonamide ≥97%

Supplier: Matrix Scientific

115192-500MG 115192-1G
102892-334EA 1243.08 USD
102892-334 102892-336
4-Chloro-N-(2,5-difluorophenyl)benzenesulfonamide ≥97%
4-Chloro-N-(2,5-difluorophenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₈ClF₂NO₂S
MDL Number: MFCD05136552
CAS Number: 290331-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR