(5,7-Dimethyl-[1,2,4]triazolo-[1,5-a]pyrimidin-2-yl)methanol ≥97%

Supplier: Matrix Scientific

Synonyms: (5,7-Dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl)methanol

115190-500MG 115190-1G
102892-278EA 624.62 USD
102892-278 102892-280
(5,7-Dimethyl-[1,2,4]triazolo-[1,5-a]pyrimidin-2-yl)methanol ≥97%
(5,7-Dimethyl-[1,2,4]triazolo-[1,5-a]pyrimidin-2-yl)methanol

Hazard: Irritant

Formula: C₈H₁₀N₄O
MW: 178.2 g/mol
MDL Number: MFCD11846959
CAS Number: 54535-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR