8-Bromo-3-iodoquinoline ≥90%

Supplier: Matrix Scientific

115189-500MG 115189-1G
102892-274EA 1243.08 USD
102892-274 102892-276
8-Bromo-3-iodoquinoline ≥90%
8-Bromo-3-iodoquinoline

Hazard: Irritant

Formula: C₉H₅BrIN
MW: 333.96 g/mol
MDL Number: MFCD09261261
CAS Number: 917251-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR