3-Cyano-2-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific

115186-500MG 115186-1G
102892-262EA 284.62 USD
102892-262 102892-264
3-Cyano-2-fluorobenzoic acid ≥97%
3-Cyano-2-fluorobenzoic acid

Hazard: Irritant

Formula: C₈H₄FNO₂
MDL Number: MFCD09261249
CAS Number: 219519-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR