4-(m-Tolylamino)pyridine-3-sulfonamide ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(m-Tolylamino)-3-sulphamoylpyridine, 4-(3-Methylphenyl)aminopyridine-3-sulphonamide, Torsemide Related Compound A, 4-(3-Methylphenyl)amino-3-sulphamoylpyridine

115185-500MG 115185-1G
102892-258EA 204.62 USD
102892-258 102892-260
4-(m-Tolylamino)pyridine-3-sulfonamide ≥97%
4-(m-Tolylamino)pyridine-3-sulfonamide

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₂S
MW: 263.32 g/mol
MDL Number: MFCD00661332
CAS Number: 72811-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR