Methyl-3-[(methylamino)sulfonyl]-1-benzothiophene-2-carboxylate ≥90%

Supplier: Matrix Scientific

115182-1G 115182-500MG
102892-248EA 1040 USD
102892-248 102895-538
Methyl-3-[(methylamino)sulfonyl]-1-benzothiophene-2-carboxylate ≥90%
Methyl-3-[(methylamino)sulfonyl]-1-benzothiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₄S₂
MW: 285.34 g/mol
MDL Number: MFCD19105040
CAS Number: 1325304-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR