N-(4-Fluorophenyl)-6-hydrazinopyridine-3-sulfonamide ≥95%

Supplier: Matrix Scientific

115178-1G 115178-500MG
102895-524EA 1040 USD
102895-524 102899-140
N-(4-Fluorophenyl)-6-hydrazinopyridine-3-sulfonamide ≥95%
N-(4-Fluorophenyl)-6-hydrazinopyridine-3-sulfonamide

Hazard: Irritant

Formula: C₁₁H₁₁FN₄O₂S
MW: 282.3 g/mol
MDL Number: MFCD12444500
CAS Number: 1040059-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR