4-[3-(3,4-Dimethylphenyl)-1,2,4-oxadiazol-5-yl]phthalazin-1(2H)-one ≥95%

Supplier: Matrix Scientific

115176-500MG 115176-1G
102899-108EA 693.85 USD
102899-108 102899-134
4-[3-(3,4-Dimethylphenyl)-1,2,4-oxadiazol-5-yl]phthalazin-1(2H)-one ≥95%
4-[3-(3,4-Dimethylphenyl)-1,2,4-oxadiazol-5-yl]phthalazin-1(2H)-one

Hazard: Irritant

Formula: C₁₈H₁₄N₄O₂
MDL Number: MFCD19105023
CAS Number: 1325306-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR