8-(Azepan-1-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one ≥95%

Supplier: Matrix Scientific

115175-500MG 115175-1G
102899-104EA 693.85 USD
102899-104 102899-106
8-(Azepan-1-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one ≥95%
8-(Azepan-1-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₃S
MDL Number: MFCD18804065
CAS Number: 1291486-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR