8-(Morpholin-4-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one ≥95%

Supplier: Matrix Scientific

115174-500MG 115174-1G
102899-100EA 693.85 USD
102899-100 102899-102
8-(Morpholin-4-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one ≥95%
8-(Morpholin-4-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

Hazard: Irritant

Formula: C₁₀H₁₂N₄O₄S
MDL Number: MFCD18804064
CAS Number: 1291486-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR