8-(Pyrrolidin-1-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one ≥95%

Supplier: Matrix Scientific

115172-500MG 115172-1G
102895-516EA 693.85 USD
102895-516 102895-518
8-(Pyrrolidin-1-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one ≥95%
8-(Pyrrolidin-1-ylsulfonyl)[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

Hazard: Irritant

Formula: C₁₀H₁₂N₄O₃S
MDL Number: MFCD18804063
CAS Number: 1291486-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR