3-(Anilinomethyl)-7-methoxyquinolin-2(1H)-one ≥95%

Supplier: Matrix Scientific

115170-500MG 115170-1G
102895-508EA 693.85 USD
102895-508 102895-510
3-(Anilinomethyl)-7-methoxyquinolin-2(1H)-one ≥95%
3-(Anilinomethyl)-7-methoxyquinolin-2(1H)-one

Hazard: Irritant

Formula: C₁₇H₁₆N₂O₂
MDL Number: MFCD11048615
CAS Number: 947018-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR