7-Methoxy-3-{[(4-methoxyphenyl)amino]methyl}quinolin-2(1H)-one ≥95%

Supplier: Matrix Scientific

115169-500MG 115169-1G
102895-504EA 693.85 USD
102895-504 102895-506
7-Methoxy-3-{[(4-methoxyphenyl)amino]methyl}quinolin-2(1H)-one ≥95%
7-Methoxy-3-{[(4-methoxyphenyl)amino]methyl}quinolin-2(1H)-one

Hazard: Irritant

Formula: C₁₈H₁₈N₂O₃
MW: 310.36 g/mol
MDL Number: MFCD02572698
CAS Number: 335222-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR