6-Oxo-5,6-dihydro[1,3]dioxolo[4,5-g]quinoline-7-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Oxo-5,6-dihydro-[1,3]dioxolo[4,5-g]quinoline-7-carbaldehyde

115168-500MG 115168-1G
102895-500EA 693.85 USD
102895-500 102895-502
6-Oxo-5,6-dihydro[1,3]dioxolo[4,5-g]quinoline-7-carbaldehyde ≥95%
6-Oxo-5,6-dihydro[1,3]dioxolo[4,5-g]quinoline-7-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₇NO₄
MW: 217.18 g/mol
MDL Number: MFCD02596873
CAS Number: 462068-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR