2-(3-Chlorophenyl)-5-(2-hydroxyethyl)-6-methylpyrimidin-4(3H)-one ≥95%

Supplier: Matrix Scientific

115163-500MG 115163-1G
102892-182EA 693.85 USD
102892-182 102892-184
2-(3-Chlorophenyl)-5-(2-hydroxyethyl)-6-methylpyrimidin-4(3H)-one ≥95%
2-(3-Chlorophenyl)-5-(2-hydroxyethyl)-6-methylpyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂O₂
MDL Number: MFCD18915344
CAS Number: 1239781-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR