2-(4-Chlorophenyl)-5-(2-hydroxyethyl)-6-methylpyrimidin-4(3H)-one ≥95%
Supplier: Matrix Scientific
115161-500MG
115161-1G
102892-174EA
693.85
USD
102892-174
102892-176
2-(4-Chlorophenyl)-5-(2-hydroxyethyl)-6-methylpyrimidin-4(3H)-one ≥95%
2-(4-Chlorophenyl)-5-(2-hydroxyethyl)-6-methylpyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₁₃H₁₃ClN₂O₂ |
MDL Number:
MFCD18915343 CAS Number: 82019-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...