2-Azido-N-1,3-benzodioxol-5-ylacetamide ≥95%
Supplier: Matrix Scientific
115160-500MG
115160-1G
102892-170EA
693.85
USD
102892-170
102892-172
2-Azido-N-1,3-benzodioxol-5-ylacetamide ≥95%
2-Azido-N-1,3-benzodioxol-5-ylacetamide
Hazard: Irritant
Formula:
C₉H₈N₄O₃ |
MDL Number:
MFCD14652716 CAS Number: 1250657-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...