2-Azido-N-(2,6-dimethylphenyl)acetamide ≥95%

Supplier: Matrix Scientific

115159-500MG 115159-1G
102892-118EA 693.85 USD
102892-118 102892-120
2-Azido-N-(2,6-dimethylphenyl)acetamide ≥95%
2-Azido-N-(2,6-dimethylphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂N₄O
MDL Number: MFCD14652502
CAS Number: 119053-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR