2-Azido-N-(2-chlorobenzyl)acetamide ≥95%

Supplier: Matrix Scientific

115158-500MG 115158-1G
102892-114EA 693.85 USD
102892-114 102892-116
2-Azido-N-(2-chlorobenzyl)acetamide ≥95%
2-Azido-N-(2-chlorobenzyl)acetamide

Hazard: Irritant

Formula: C₉H₉ClN₄O
MDL Number: MFCD14652518
CAS Number: 1249051-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR