2-Azido-N-(5-chloro-2-methoxyphenyl)acetamide ≥95%

Supplier: Matrix Scientific

115154-500MG 115154-1G
102891-912EA 693.85 USD
102891-912 102891-914
2-Azido-N-(5-chloro-2-methoxyphenyl)acetamide ≥95%
2-Azido-N-(5-chloro-2-methoxyphenyl)acetamide

Hazard: Irritant

Formula: C₉H₉ClN₄O₂
MDL Number: MFCD14652679
CAS Number: 1160748-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR