Ethyl-4-[(azidoacetyl)amino]benzoate ≥95%

Supplier: Matrix Scientific

115153-500MG 115153-1G
102891-908EA 693.85 USD
102891-908 102891-910
Ethyl-4-[(azidoacetyl)amino]benzoate ≥95%
Ethyl-4-[(azidoacetyl)amino]benzoate

Hazard: Irritant

Formula: C₁₁H₁₂N₄O₃
MDL Number: MFCD18185036
CAS Number: 1160748-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR