2-Azido-N-(4-methylphenyl)acetamide ≥95%

Supplier: Matrix Scientific

115152-500MG 115152-1G
102891-904EA 693.85 USD
102891-904 102891-906
2-Azido-N-(4-methylphenyl)acetamide ≥95%
2-Azido-N-(4-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₁₀N₄O
MDL Number: MFCD14652267
CAS Number: 116433-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR