2-Azido-N-(3,4-dimethylphenyl)acetamide ≥95%

Supplier: Matrix Scientific

115148-500MG 115148-1G
102891-840EA 693.85 USD
102891-840 102891-842
2-Azido-N-(3,4-dimethylphenyl)acetamide ≥95%
2-Azido-N-(3,4-dimethylphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂N₄O
MDL Number: MFCD14652500
CAS Number: 1160748-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR