4-Aminobenzamidine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Aminophenylamidine dihydrochloride, 4-Aminobenzene-1-carboximidamide dihydrochloride

115147-500MG 115147-1G
102891-836EA 693.85 USD
102891-836 102891-838
4-Aminobenzamidine dihydrochloride ≥95%
4-Aminobenzamidine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₁Cl₂N₃
MW: 208.09 g/mol
MDL Number: MFCD00013001
CAS Number: 2498-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR