Ethyl L-valinate ≥95%

Supplier: Matrix Scientific

115142-500MG 115142-1G
102891-828EA 693.85 USD
102891-828 102891-830
Ethyl L-valinate ≥95%
Ethyl L-valinate

Hazard: Irritant

Formula: C₇H₁₅NO₂
MDL Number: MFCD12796096
CAS Number: 17431-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR