2-(Chloromethyl)-6-fluoroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

115141-500MG 115141-1G
102895-488EA 693.85 USD
102895-488 102895-490
2-(Chloromethyl)-6-fluoroimidazo[1,2-a]pyridine ≥95%
2-(Chloromethyl)-6-fluoroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₆ClFN₂
MDL Number: MFCD09994962
CAS Number: 872363-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR