4-Oxo-6-phenyl-4,5-dihydropyrazolo[1,5-a]pyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

115131-1G 115131-500MG
102891-756EA 1040 USD
102891-756 102895-472
4-Oxo-6-phenyl-4,5-dihydropyrazolo[1,5-a]pyrazine-2-carboxylic acid ≥95%
4-Oxo-6-phenyl-4,5-dihydropyrazolo[1,5-a]pyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉N₃O₃
MDL Number: MFCD25953192
CAS Number: 1443978-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR