3-(3-Pyridin-4-yl-1,2,4-oxadiazol-5-yl)pyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

115126-500MG 115126-1G
102899-082EA 693.85 USD
102899-082 102899-084
3-(3-Pyridin-4-yl-1,2,4-oxadiazol-5-yl)pyridin-2(1H)-one ≥95%
3-(3-Pyridin-4-yl-1,2,4-oxadiazol-5-yl)pyridin-2(1H)-one

Hazard: Irritant

Formula: C₁₂H₈N₄O₂
MDL Number: MFCD16875009
CAS Number: 1239787-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR