2-(2-Azidoethyl)-1H-isoindole-1,3(2H)-dione ≥95%

Supplier: Matrix Scientific

115123-500MG 115123-1G
102899-070EA 693.85 USD
102899-070 102899-072
2-(2-Azidoethyl)-1H-isoindole-1,3(2H)-dione ≥95%
2-(2-Azidoethyl)-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₀H₈N₄O₂
MDL Number: MFCD14644895
CAS Number: 30250-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR