4-[3-(2-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride ≥95%

Supplier: Matrix Scientific

115121-500MG 115121-1G
102898-984EA 693.85 USD
102898-984 102898-986
4-[3-(2-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride ≥95%
4-[3-(2-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClN₃O₂
MDL Number: MFCD27960949
CAS Number: 1018626-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR